Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate

A101267-100MG A101267-250MG A101267-5G
103795-880EA 50 USD
103795-880 103821-566 76717-522 76717-524 77297-034
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-bromo-1-methyl-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈BrNO₂
MW: 218.06 g/mol
Melting Pt: 63…65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275077
CAS Number: 1196-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR