Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1-methyl-1H-pyrazole-3-carboxylate

A133932-100MG A133932-250MG A133932-25G A133932-5G A133932-1G
103801-936EA 21 USD
103801-936 77027-198 77027-200 77298-666 77298-668 77314-636
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate 97%
Methyl-4-bromo-1-methyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
Boiling Pt: 276.5 °C (760 mm Hg)
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02253785
CAS Number: 211738-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR