Methyl 4-bromo-1-methyl-1H-imidazole-2-carboxylate 95%

Supplier: Ambeed
Warning

A112424-10G A112424-250MG A112424-1G A112424-5G A112424-25G
103798-008EA 28 USD
103798-008 103823-842 76809-990 76809-992 77284-240 77284-242 77882-937
Methyl 4-bromo-1-methyl-1H-imidazole-2-carboxylate 95%
Methyl 4-bromo-1-methyl-1H-imidazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13189816
CAS Number: 864076-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR