Methyl-4-bromo-1H-pyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

A106399-10G A106399-25G A106399-1G A106399-5G A106399-100G
103797-030EA 5 USD
103797-030 103822-784 76869-458 76869-460 77309-638 77309-640 77549-044
Methyl-4-bromo-1H-pyrrole-2-carboxylate 97%
Methyl-4-bromo-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂
Boiling Pt: 290.5 °C (760 mm Hg)
Melting Pt: 100…101 °C
Flash Pt: 129.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00832855
CAS Number: 934-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR