Methyl-4-bromo-1H-pyrazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A417464-1G A417464-25G A417464-5G A417464-100G A417464-10G A417464-250MG A417464-500G
103827-332EA 33 USD
103827-332 103838-378 76841-960 76841-962 76841-964 77033-824 77033-826 77033-828 77033-830
Methyl-4-bromo-1H-pyrazole-3-carboxylate 95%
Methyl-4-bromo-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
Boiling Pt: 318 °C (760 mm Hg)
Flash Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00466326
CAS Number: 81190-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR