Methyl-4-bromoindole-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1H-indole-6-carboxylate

A101336-100MG A101336-10G A101336-250MG A101336-25G A101336-1G A101336-5G
103795-914EA 14 USD
103795-914 103821-600 76821-232 76821-234 76821-236 76821-238 77282-702 77282-704
Methyl-4-bromoindole-6-carboxylate 97%
Methyl-4-bromoindole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03867040
CAS Number: 882679-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR