Methyl-4-bromo-6-(1H)-indazole carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-bromo-1H-indazole-6-carboxylate

A104584-250MG A104584-1G A104584-5G A104584-25G
103796-736EA 15 USD
103796-736 103822-456 76821-454 77309-314 77309-316 77682-962
Methyl-4-bromo-6-(1H)-indazole carboxylate 97%
Methyl-4-bromo-6-(1H)-indazole carboxylate

Precaución: For research use only.

Formula: C₉H₇BrN₂O₂
MW: 255.08 g/mol
Boiling Pt: 395.9±22.0 °C (760 mm Hg)
Flash Pt: 193.2 °C
Storage Temperature: Ambient
MDL Number: MFCD07368212
CAS Number: 885518-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR