Methyl-4-aminothiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-aminothiophene-3-carboxylate

A202435-5G A202435-100MG A202435-250MG A202435-1G
103817-388EA 160 USD
103817-388 103848-028 77086-140 77112-200 77112-202 77302-812
Methyl-4-aminothiophene-3-carboxylate 95%
Methyl-4-aminothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Boiling Pt: 295.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00130102
CAS Number: 69363-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR