Methyl-4-aminothiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-thiophene-2-carboxylic acid methyl ester, Methyl 4-aminothiophene-2-carboxylate

A395419-5G A395419-250MG
103826-962EA 48 USD
103826-962 103838-000 76841-420 77291-112
Methyl-4-aminothiophene-2-carboxylate 98%
Methyl-4-aminothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
Melting Pt: 82…84 °C
Flash Pt: 146.677 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368326
CAS Number: 89499-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR