Methyl 4-amino-6-chloronicotinate 95%

Supplier: Ambeed
Warning

A243954-100MG A243954-10G A243954-250MG A243954-1G A243954-5G
103811-556EA 78 USD
103811-556 103851-380 76874-480 76874-482 76874-484 77090-586 77090-588
Methyl 4-amino-6-chloronicotinate 95%
Methyl 4-amino-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18259038
CAS Number: 1256785-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR