Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-5-iodo-2-methoxybenzoate

A228259-100MG A228259-250MG A228259-1G A228259-10G
103819-380EA 44 USD
103819-380 103850-288 76976-022 76976-024 77089-074 77881-584
Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate 97%
Methyl-4-amino-5-iodo-2-methoxybenzenecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀INO₃
Storage Temperature: Refrigerator
MDL Number: MFCD11112895
CAS Number: 137832-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR