Methyl-4-amino-3-nitrobenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-amino-3-nitrobenzoate
A390084-10G
A390084-250MG
A390084-1G
A390084-25G
A390084-5G
A390084-100G
103826-860EA
6
USD
103826-860
103837-902
76724-956
76724-958
76841-284
76841-286
76841-288
77668-798
Methyl-4-amino-3-nitrobenzoate 95%
Methyl 4-amino-3-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈N₂O₄ MW: 196.16 g/mol Boiling Pt: 392.8 °C (760 mm Hg) Melting Pt: 204…206 °C Flash Pt: 191.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017562 CAS Number: 3987-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...