Methyl 4-amino-3-methyl-1H-pyrazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A131727-100MG A131727-250MG
103801-620EA 444 USD
103801-620 77234-562 77234-564
Methyl 4-amino-3-methyl-1H-pyrazole-5-carboxylate 97%
Methyl 4-amino-3-methyl-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.16 g/mol
Boiling Pt: 353.4±37.0 °C (760 mm Hg)
Flash Pt: 167.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD21601716
CAS Number: 1587700-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR