Methyl-4-amino-3-methoxybenzoate 98%

Supplier: Ambeed
Warning

A244388-100G A244388-10G A244388-1G A244388-25G A244388-5G
103811-578EA 5 USD
103811-578 103851-410 76692-344 76692-346 77090-616 77090-618 77090-620
Methyl-4-amino-3-methoxybenzoate 98%
Methyl 4-amino-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
Boiling Pt: 332 °C (760 mm Hg)
Melting Pt: 128…131 °C
Flash Pt: 175.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017203
CAS Number: 41608-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR