Methyl-4-amino-3-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-amino-3-fluorobenzoate
A142432-10G
A142432-1G
A142432-25G
A142432-5G
103803-158EA
5
USD
103803-158
76996-910
77299-290
77316-386
77316-388
Methyl-4-amino-3-fluorobenzoate 98%
Methyl 4-amino-3-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈FNO₂ MW: 169.1530 g/mol Melting Pt: 109…110 °C Flash Pt: 133.016 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689703 CAS Number: 185629-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...