Methyl-4-amino-3-bromobenzoate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 4-amino-3-bromobenzoate
A432957-25G
A432957-5G
A432957-10G
A432957-1G
103827-540EA
9
USD
103827-540
103838-606
76842-308
76842-310
77004-210
77307-260
Methyl-4-amino-3-bromobenzoate 98%
Methyl 4-amino-3-bromobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol Melting Pt: 105…109 °C Flash Pt: 155.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861385 CAS Number: 106896-49-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...