Methyl 4-amino-3,5-dimethylbenzoate 97%

Supplier: Ambeed
Warning

A291266-1G A291266-25G A291266-5G A291266-250MG
103813-904EA 16 USD
103813-904 103835-044 76837-634 76837-636 76837-638 77289-796
Methyl 4-amino-3,5-dimethylbenzoate 97%
Methyl 4-amino-3,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Boiling Pt: 331.3 °C (760 mm Hg)
Flash Pt: 180.604 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09833906
CAS Number: 3095-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR