Methyl 4-amino-3-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Warning

A180703-100MG A180703-250MG A180703-1G
103807-552EA 46 USD
103807-552 103845-912 76813-542 76813-544 77301-688
Methyl 4-amino-3-(trifluoromethyl)benzoate 98%
Methyl 4-amino-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.161 g/mol
Boiling Pt: 305.7 °C (760 mm Hg)
Flash Pt: 138.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11226326
CAS Number: 167760-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR