Methyl 4-amino-3-(tert-butyl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

A101481-100MG A101481-250MG A101481-1G
103795-996EA 107 USD
103795-996 103821-680 77245-136 77245-138 77297-122
Methyl 4-amino-3-(tert-butyl)benzoate hydrochloride 97%
Methyl 4-amino-3-(tert-butyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₈ClNO₂
MW: 243.73 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30609550
CAS Number: 2044702-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR