Methyl-4-amino-2-methylbenzoate 98%
Supplier: Ambeed
|
Warning
|
A278053-25G
A278053-5G
A278053-100G
A278053-10G
103813-388EA
11
USD
103813-388
103853-326
77093-234
77093-236
77276-898
77276-900
77305-286
Methyl-4-amino-2-methylbenzoate 98%
Methyl 4-amino-2-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₂ Boiling Pt: 302.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08458829 CAS Number: 6933-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...