Methyl 4-amino-2-bromobenzoate 95%

Supplier: Ambeed
Warning

A131604-100G A131604-100MG A131604-10G A131604-250MG A131604-1G A131604-25G A131604-5G
103801-608EA 19 USD
103801-608 76983-732 76983-734 76983-736 76983-738 77314-106 77314-108 77314-110
Methyl 4-amino-2-bromobenzoate 95%
Methyl 4-amino-2-bromobenzoate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Boiling Pt: 345.2 °C (760 mm Hg)
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00859376
CAS Number: 98545-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR