Methyl 4-amino-2,6-dimethylbenzoate 97%

Supplier: Ambeed
Warning

A177972-250MG A177972-1G
103799-434EA 46 USD
103799-434 103807-284 103845-624 77011-854 77301-534
Methyl 4-amino-2,6-dimethylbenzoate 97%
Methyl 4-amino-2,6-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20691178
CAS Number: 79909-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR