Methyl-4-amino-2,6-difluorobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-2,6-difluorobenzoate

A166456-100MG A166456-250MG A166456-1G
103806-070EA 52 USD
103806-070 103844-332 76813-028 76813-030 77320-582
Methyl-4-amino-2,6-difluorobenzoate 97%
Methyl-4-amino-2,6-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MW: 187.14 g/mol
Boiling Pt: 306 °C (760 mm Hg)
Flash Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16038807
CAS Number: 191478-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR