Methyl 4,6-dichloropyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

A171695-100MG A171695-250MG A171695-1G A171695-5G
103806-658EA 38 USD
103806-658 103844-934 77011-600 77011-602 77082-106 77301-144
Methyl 4,6-dichloropyrimidine-5-carboxylate 95%
Methyl 4,6-dichloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MW: 207.01 g/mol
Melting Pt: 47…49 °C
Storage Temperature: Freezer
MDL Number: MFCD10698883
CAS Number: 87600-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR