Methyl-4,6-dichloropyridazine-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4,6-dichloropyridazine-3-carboxylate

A220117-10G A220117-250MG A220117-1G A220117-100G
103818-836EA 8 USD
103818-836 103849-718 76785-940 76785-942 77088-310 77568-126
Methyl-4,6-dichloropyridazine-3-carboxylate 98%
Methyl-4,6-dichloropyridazine-3-carboxylate

Precaución: For research use only.

Formula: C₆H₄Cl₂N₂O₂
MW: 207.0142 g/mol
Boiling Pt: 345.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09953612
CAS Number: 372118-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR