Methyl-4,6-dichloropicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4,6-dichloropyridine-2-carboxylate, Methyl 4,6-dichloropicolinate

A102854-10G A102854-250MG A102854-25G A102854-1G A102854-5G
103796-432EA 30 USD
103796-432 103822-130 76970-248 76970-250 76970-252 77308-998 77309-000
Methyl-4,6-dichloropicolinate 98%
Methyl 4,6-dichloropicolinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.0261 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12965121
CAS Number: 98273-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR