Methyl-4,6-dichloronicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4,6-dichloronicotinate
A178839-100G
A178839-10G
A178839-25G
A178839-5G
A178839-1G
103807-374EA
5
USD
103807-374
103845-714
76813-490
76813-492
77083-156
77083-158
77301-576
Methyl-4,6-dichloronicotinate 98%
Methyl-4,6-dichloronicotinate
Precaución: for Research Use Only
Formula:
C₇H₅Cl₂NO₂ MW: 206.03 g/mol Boiling Pt: 260 °C (760 mm Hg) Melting Pt: 41…43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04125732 CAS Number: 65973-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...