Methyl 4,6-dibromo-3-hydroxypicolinate 97%

Supplier: Ambeed
Warning

A148935-100MG A148935-1G
103803-984EA 286 USD
103803-984 76823-768 77317-574
Methyl 4,6-dibromo-3-hydroxypicolinate 97%
Methyl 4,6-dibromo-3-hydroxypicolinate

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₃
MW: 310.93 g/mol
Boiling Pt: 389.3±37.0 °C (760 mm Hg)
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD19440848
CAS Number: 321596-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR