Methyl 4,5-diaminopicolinate 95%

Supplier: Ambeed
Warning

A135632-100MG A135632-250MG
103802-186EA 201 USD
103802-186 76823-208 77611-220
Methyl 4,5-diaminopicolinate 95%
Methyl 4,5-diaminopicolinate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD13186746
CAS Number: 850689-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR