Methyl 4-(piperidin-4-yl)benzoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A147470-100MG
A147470-250MG
A147470-1G
103803-782EA
416
USD
103803-782
103841-884
77027-960
77027-962
77317-260
Methyl 4-(piperidin-4-yl)benzoate hydrochloride 97%
Methyl 4-(piperidin-4-yl)benzoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₂·ClH MW: 255.741 g/mol Boiling Pt: 371.4 °C (760 mm Hg) Flash Pt: 178.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02179141 CAS Number: 936130-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...