Methyl 4-(N-(tert-butoxycarbonyl)carbamimidoyl)benzoate 95%

Supplier: Ambeed
Warning

A201593-10G A201593-1G A201593-5G
103817-308EA 58 USD
103817-308 103847-948 77012-652 77086-026 77086-028
Methyl 4-(N-(tert-butoxycarbonyl)carbamimidoyl)benzoate 95%
Methyl 4-(N-(tert-butoxycarbonyl)carbamimidoyl)benzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₄
MW: 278.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20261468
CAS Number: 135321-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR