Methyl 4-(methylamino)butanoate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-(methylamino)butanoate hcl

A235971-1G A235971-5G A235971-100MG A235971-250MG
103811-056EA 15 USD
103811-056 103850-826 77089-856 77089-858 77153-002 77153-004
Methyl 4-(methylamino)butanoate hydrochloride 95%
Methyl 4-(methylamino)butanoate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11505402
CAS Number: 89584-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR