Methyl-4-(methylamino)-3-nitrobenzenecarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(methylamino)-3-nitrobenzoate
A153631-10G
A153631-250MG
A153631-1G
A153631-25G
103804-540EA
27
USD
103804-540
103842-716
76997-436
76997-438
77318-380
77318-382
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate 97%
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₀N₂O₄ MW: 210.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09862450 CAS Number: 36242-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...