Methyl-4-(chloromethyl)benzoate 98%

Supplier: Ambeed
Danger

A126526-10G A126526-250MG A126526-5G
76718-742EA 11 USD
76718-742 76718-744 76718-746
Methyl-4-(chloromethyl)benzoate 98%
Methyl 4-(chloromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Boiling Pt: 275.7 °C (760 mm Hg)
Melting Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00511224
CAS Number: 34040-64-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR