Methyl-4-(bromomethyl)-3-methoxybenzoate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 4-(bromomethyl)-3-methoxybenzoate
A779374-10G
A779374-1G
A779374-25G
A779374-5G
103840-972EA
24
USD
103840-972
76652-996
76845-232
76845-234
77308-312
Methyl-4-(bromomethyl)-3-methoxybenzoate 98%
Methyl 4-(bromomethyl)-3-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₃ MW: 259.0965 g/mol Melting Pt: 92…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00270115 CAS Number: 70264-94-7 UN: 3261 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...