Methyl 4-(bromomethyl)-3-fluorobenzoate 95%

Supplier: Ambeed
Danger

A211185-1G A211185-100G A211185-100MG A211185-10G A211185-250MG A211185-25G
103818-088EA 22 USD
103818-088 103848-782 77087-190 77287-954 77287-956 77287-958 77287-960 77287-962
Methyl 4-(bromomethyl)-3-fluorobenzoate 95%
Methyl 4-(bromomethyl)-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00273345
CAS Number: 128577-47-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR