Methyl 4-(bromomethyl)-2-fluorobenzoate 95%

Supplier: Ambeed
Danger

A212033-100MG A212033-250MG A212033-1G
103818-160EA 273 USD
103818-160 103848-856 77164-974 77164-976 77303-214
Methyl 4-(bromomethyl)-2-fluorobenzoate 95%
Methyl 4-(bromomethyl)-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11976782
CAS Number: 85070-57-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR