Methyl 4-(bromomethyl)-2,5-difluorobenzoate 97%

Supplier: Ambeed
Danger

A178037-100MG A178037-250MG A178037-1G A178037-5G
103807-294EA 216 USD
103807-294 103845-634 76647-260 76647-262 77301-536 77301-538
Methyl 4-(bromomethyl)-2,5-difluorobenzoate 97%
Methyl 4-(bromomethyl)-2,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD27959326
CAS Number: 1355488-72-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR