Methyl 4-(benzyloxy)-3-methoxybenzoate 98%

Supplier: Ambeed
Warning

A170387-100G A170387-10G A170387-1G A170387-25G A170387-5G
103806-480EA 11 USD
103806-480 103844-756 76720-494 76720-496 77081-866 77081-868 77081-870
Methyl 4-(benzyloxy)-3-methoxybenzoate 98%
Methyl 4-(benzyloxy)-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₄
MW: 272.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017194
CAS Number: 56441-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR