Methyl 4-(benzyloxy)-3-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A168382-100MG A168382-250MG A168382-5G A168382-1G
103806-288EA 37 USD
103806-288 76824-404 76824-406 76824-408 77081-600
Methyl 4-(benzyloxy)-3-hydroxybenzoate 95%
Methyl 4-(benzyloxy)-3-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD16987710
CAS Number: 87687-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR