Methyl-4-(aminomethyl)benzoate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)benzoic Acid Methyl Ester Hydrochloride, Methyl 4-(aminomethyl)benzoate hydrochloride

A160354-100G A160354-10G A160354-500G A160354-1G A160354-25G A160354-5G
103805-370EA 6 USD
103805-370 103843-598 77096-126 77096-128 77096-130 77300-458 77319-528 77319-530
Methyl-4-(aminomethyl)benzoate hydrochloride 97%
Methyl-4-(aminomethyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 201.65 g/mol
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00182671
CAS Number: 6232-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR