4-(Methoxycarbonyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

A101074-100G A101074-25G A101074-500G A101074-1G A101074-5G
103795-792EA 5 USD
103795-792 103821-482 76780-448 76780-450 76780-452 77296-978 77296-980
4-(Methoxycarbonyl)phenylboronic acid pinacol ester 98%
4-(Methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MW: 262.11 g/mol
Boiling Pt: 355.6 °C (760 mm Hg)
Melting Pt: 77…81 °C
Flash Pt: 194.3 °C
Storage Temperature: Ambient
MDL Number: MFCD02179438
CAS Number: 171364-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR