Methyl 4-(2-hydroxyphenyl)butanoate 95%
Supplier: Ambeed
|
Warning
|
A437501-100MG
A437501-10G
A437501-250MG
A437501-1G
A437501-5G
103827-628EA
35
USD
103827-628
103838-694
77264-948
77264-950
77264-952
77307-296
77307-298
Methyl 4-(2-hydroxyphenyl)butanoate 95%
Methyl 4-(2-hydroxyphenyl)butanoate
Precaución: Inert atmosphere, room temperature
Formula:
C₁₁H₁₄O₃ MW: 194.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24714538 CAS Number: 93108-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...