Methyl-4-(2-bromoethyl)benzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-(2-bromoethyl)benzoate
A213242-10G
A213242-250MG
A213242-1G
A213242-25G
A213242-5G
103818-276EA
20
USD
103818-276
103848-982
76825-958
76825-960
77087-478
77087-480
77087-482
Methyl-4-(2-bromoethyl)benzoate 97%
Methyl-4-(2-bromoethyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₂ MW: 243.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14708225 CAS Number: 136333-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...