Methyl 4-(1-bromoethyl)benzoate 97%

Supplier: Ambeed
Danger

A120437-250MG A120437-1G A120437-5G
103799-932EA 47 USD
103799-932 77161-276 77312-036 77312-038
Methyl 4-(1-bromoethyl)benzoate 97%
Methyl 4-(1-bromoethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18452043
CAS Number: 16281-97-3
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR