Methyl 4-(1-aminocyclopropyl)benzoate hydrochloride 95%

Supplier: Ambeed
Warning

A479131-1G A479131-100MG A479131-250MG
103828-110EA 107 USD
103828-110 103839-182 76843-120 76979-732 76979-734
Methyl 4-(1-aminocyclopropyl)benzoate hydrochloride 95%
Methyl 4-(1-aminocyclopropyl)benzoate hydrochloride

Precaución: For research use only

Formula: C₁₁H₁₄ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11616336
CAS Number: 1014645-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR