4-(Methoxycarbonyl)benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl)benzoate, Methyl 4-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]benzoate

A171844-100MG A171844-250MG A171844-1G A171844-5G
103806-668EA 108 USD
103806-668 103844-948 77151-292 77151-294 77301-164 77301-166
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester 97%
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MW: 276.136 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383373
CAS Number: 150033-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR