Methyl 4-(((2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl)amino)-2-methoxybenzoate 97%

Supplier: Ambeed
Warning

A100807-250MG A100807-1G A100807-5G
103795-638EA 17 USD
103795-638 103821-330 77245-082 77296-846 77296-848
Methyl 4-(((2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl)amino)-2-methoxybenzoate 97%
Methyl 4-(((2,2-dimethyl-4,6-dioxo-1,3-dioxan-5-ylidene)methyl)amino)-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₆H₁₇NO₇
MW: 335.3 g/mol
Boiling Pt: 547.8±50.0 °C (760 mm Hg)
Flash Pt: 285.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26395147
CAS Number: 205448-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR