Methyl-3-phenylpropionate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl dihydrocinnamate, 3-Phenylpropionic acid methyl ester, Methyl 3-phenylpropionate, Hydrocinnamic acid methyl ester

A254663-100G A254663-10G A254663-500G A254663-1000G A254663-25G
103809-810EA 175 USD
103809-810 103852-004 77031-632 77031-634 77031-636 77091-440 77091-442
Methyl-3-phenylpropionate 98%
Methyl-3-phenylpropionate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Boiling Pt: 238.5 °C (760 mm Hg)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00017209
CAS Number: 103-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR