Methyl butyrylacetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 3-oxohexanoate
A217235-1000G
A217235-100G
A217235-10G
A217235-500G
A217235-25G
A217235-5G
103849-340EA
5
USD
103849-340
76721-854
76721-856
76721-858
76721-860
77087-956
77303-448
Methyl butyrylacetate 98%
Methyl butyrylacetate
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ Boiling Pt: 193 °C (760 mm Hg) Melting Pt: 43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040995 CAS Number: 30414-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...